Learn More
Lonza Walkersville TheraPEAK X-VIVO 10 SF Med
Catalog No. 5000496457
Encompass
TheraPEAK X-VIVO 10 Media formulations are designed to support slower growing less mature cells such as hematopoetic stem cells and natural killer cells TheraPEAK X-VIVO 10 Media also supports the generation of Lymphokine activated killer (LAK) cells and peripheral blood lymphocytes (PBL) TheraPEAK X-VIVO 10 Media is available as a 1X liquid in 3 convenient formulations as off the shelf catalog items Additional customization of packaging available X-VIVO 10 Media catalog number (BE)04-380Q contains L-glutamine gentamicin and phenol red and is only available as research use only TheraPEAK X-VIVO 10 catalog number (BE)BP04-743Q contains L-glutamine but does not contain gentamicin or phenol red TheraPEAK X-VIVO 10 catalog number BE02-055Q contains L-glutamine and recombinant transferrin but does not contain gentamicin or phenol red The recombinant transferrin replaces native human transferrin
Catalog No. 50-004-96457 Supplier Lonza Walkersville Supplier No. BP04743Q
TheraPEAK X-VIVO 10 Media formulations are designed to support slower growing less mature cells such as hematopoetic stem cells and natural killer cells TheraPEAK X-VIVO 10 Media also supports the generation of Lymphokine activated killer (LAK) cells and peripheral blood lymphocytes (PBL) TheraPEAK X-VIVO 10 Media is available as a 1X liquid in 3 convenient formulations as off the shelf catalog items Additional customization of packaging available X-VIVO 10 Media catalog number (BE)04-380Q contains L-glutamine gentamicin and phenol red and is only available as research use only TheraPEAK X-VIVO 10 catalog number (BE)BP04-743Q contains L-glutamine but does not contain gentamicin or phenol red TheraPEAK X-VIVO 10 catalog number BE02-055Q contains L-glutamine and recombinant transferrin but does not contain gentamicin or phenol red The recombinant transferrin replaces native human transferrin