Learn More
Ace Glass, Inc. Thermometer, 0 to 230°F, 12in length, 76mm immersion, accuracy ±2°F
SDP
Catalog No. 50974535
Encompass
76mm immersion thermometer ranging from 0 to 230F. Thermometer is 12 in length and has an accuracy of 2F. In case of breakage this PFA coated thermometer will contain the organic black liquid filling. Thermometer comes with a NIST traceable statement of accuracy.
Catalog No. 50-974-535 Supplier Ace Glass, Inc. Supplier No. 828315
76mm immersion thermometer ranging from 0 to 230F. Thermometer is 12 in length and has an accuracy of 2F. In case of breakage this PFA coated thermometer will contain the organic black liquid filling. Thermometer comes with a NIST traceable statement of accuracy.