Learn More
Ace Glass, Inc. Thermometer, 0 to 300°F, 12" length, total immersion
SDP
Catalog No. 50974548
Encompass
0 to 300F, total immersion, enviro-safe economy thermometer. 12 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.
Catalog No. 50-974-548 Supplier Ace Glass, Inc. Supplier No. 829438
0 to 300F, total immersion, enviro-safe economy thermometer. 12 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.