Learn More
Thermco Products Inc THERM -1+51C ORGNC TOTAL IMMER
Catalog No. NC1058135
Encompass_Preferred
Non-mercury thermometer made to the specifications of American National Standards Institute. Accurate to N.I.S.T. Supplied with a statement of traceability, individually serialized. Eligible to be certified at individual temperature points and used as N.I.S.T. standard to verify other temperature instruments.
Catalog No. NC1058135 Supplier Thermco Products Inc Supplier No. ACC1003S
Non-mercury thermometer made to the specifications of American National Standards Institute. Accurate to N.I.S.T. Supplied with a statement of traceability, individually serialized. Eligible to be certified at individual temperature points and used as N.I.S.T. standard to verify other temperature instruments.