Learn More
Ace Glass, Inc. Thermometer, -10 to 150°C, 8" length, 50mm immersion
SDP
Catalog No. 50974543
Encompass
-10 to 150C enviro-safe economy thermometer with a 50mm immersion length. 8 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.
Catalog No. 50-974-543 Supplier Ace Glass, Inc. Supplier No. 829406
-10 to 150C enviro-safe economy thermometer with a 50mm immersion length. 8 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.