Learn More
Ace Glass, Inc. Thermometer, -10 to 150°C, 8in length, 50mm immersion, accuracy ±1°C, ±1.5°C above 110°C
SDP
Catalog No. 50974532
Encompass
50mm immersion thermometer ranging from -10 to 150C. Thermometer is 8 in length and has an accuracy of 1C, 1.5C above 110C. In case of breakage this PFA coated thermometer will contain the organic black liquid filling. Thermometer comes with a NIST traceable statement of accuracy.
Catalog No. 50-974-532 Supplier Ace Glass, Inc. Supplier No. 828303
50mm immersion thermometer ranging from -10 to 150C. Thermometer is 8 in length and has an accuracy of 1C, 1.5C above 110C. In case of breakage this PFA coated thermometer will contain the organic black liquid filling. Thermometer comes with a NIST traceable statement of accuracy.