Learn More
Ace Glass, Inc. 25mm non-mercury immersion thermometer
SDP
Catalog No. 50978365
Encompass_Preferred
25mm non-mercury immersion thermometer for use with specific gravity bottles. Thermometer has a range of 14 to 37 degrees C with 0.2 subdivisions, and a 10/18 standard taper joint.
Catalog No. 50-978-365 Supplier Ace Glass, Inc. Supplier No. 541506
25mm non-mercury immersion thermometer for use with specific gravity bottles. Thermometer has a range of 14 to 37 degrees C with 0.2 subdivisions, and a 10/18 standard taper joint.