Learn More
Ace Glass, Inc. Thermometer, -10 to 200°C, 12" length, 76mm immersion
SDP
Catalog No. 50974544
Encompass
-10 to 200C enviro-safe economy thermometer with a 76mm immersion length. 12 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.
Catalog No. 50-974-544 Supplier Ace Glass, Inc. Supplier No. 829414
-10 to 200C enviro-safe economy thermometer with a 76mm immersion length. 12 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.