Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ace Glass, Inc. Thermometer, -100 to 50c, 12" length, 76mm immersion
SDP

Supplier:  Ace Glass, Inc. 829306

Encompass

Thermometer, -100 to 50c, 12" length, 76mm immersion

Catalog No. 50-955-8448


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.