Learn More
Lauda Brinkmann Lp THERM 160 BATH LIQUID 10 L DRU
Catalog No. 50931355
Encompass_Preferred
Bath Liquid Therm 160 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 60 to 160oC, 10 L Drum This item is non returnable*
Catalog No. 50-931-355 Supplier Lauda Brinkmann Lp Supplier No. LZB 206
Bath Liquid Therm 160 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 60 to 160oC, 10 L Drum This item is non returnable*