Learn More
Lauda Brinkmann Lp THERM 160 BATH LIQUID 20L DRUM
Catalog No. 50931356
Encompass_Preferred
Bath Liquid Therm 160 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 60 to 160oC, 20 L Drum This item is non returnable*
Catalog No. 50-931-356 Supplier Lauda Brinkmann Lp Supplier No. LZB 306
Bath Liquid Therm 160 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 60 to 160oC, 20 L Drum This item is non returnable*