Learn More
Ace Glass, Inc. Thermometer, -20 to 110°C, 12" length, 76mm immersion
SDP
Catalog No. 50974541
Encompass
-20 to 110C enviro-safe economy thermometer with a 76mm immersion length. 12 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.
Catalog No. 50-974-541 Supplier Ace Glass, Inc. Supplier No. 829404
-20 to 110C enviro-safe economy thermometer with a 76mm immersion length. 12 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.