Learn More
Ace Glass, Inc. Thermometer, -20 to 110°C, 12" length, total immersion
SDP
Catalog No. 50974542
Encompass
-20 to 110C, total immersion, enviro-safe economy thermometer. 12 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.
Catalog No. 50-974-542 Supplier Ace Glass, Inc. Supplier No. 829405
-20 to 110C, total immersion, enviro-safe economy thermometer. 12 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.