Learn More
Ace Glass, Inc. Thermometer, -20 to 150°C, 12" length, total immersion(organic blue spirit)
SDP
Catalog No. 50979764
Encompass
-20 to 150C, total immersion, enviro-safe economy thermometer. 12 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.
Catalog No. 50-979-764 Supplier Ace Glass, Inc. Supplier No. 829411
-20 to 150C, total immersion, enviro-safe economy thermometer. 12 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.