Learn More
Ace Glass, Inc. Thermometer, -20 to 150°C, 12in length, total immersion, accuracy ±1°C, ±1.5°C above 110°C
SDP
Catalog No. 50979761
Encompass
Thermometer, -20 to 150°C, 12in length, total immersion, accuracy ±1°C, ±1.5°C above 110°C
Catalog No. 50-979-761 Supplier Ace Glass, Inc. Supplier No. 828311
Thermometer, -20 to 150°C, 12in length, total immersion, accuracy ±1°C, ±1.5°C above 110°C