Learn More
Ace Glass, Inc. Thermometer, 20 to 300°F, 8" length, 50mm immersion
SDP
Catalog No. 50974546
Encompass
20 to 300F enviro-safe economy thermometer with a 50mm immersion length. 8 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.
Catalog No. 50-974-546 Supplier Ace Glass, Inc. Supplier No. 829416
20 to 300F enviro-safe economy thermometer with a 50mm immersion length. 8 inches long. Thermometer is filled with organic black liquid, and is made from lead-free glass. Does not contain mercury. Comes with a NIST traceable statement of accuracy.