Learn More
Lauda Brinkmann Lp THERM 240 BATH LIQUID 10 L DRU
Catalog No. 50931364
Encompass_Preferred
Bath Liquid Therm 240 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 50 to 240oC, 10 L Drum This item is non returnable*
Catalog No. 50-931-364 Supplier Lauda Brinkmann Lp Supplier No. LZB 222
Bath Liquid Therm 240 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 50 to 240oC, 10 L Drum This item is non returnable*