Learn More
Lauda Brinkmann Lp THERM 240 BATH LIQUID 20L DRUM
Catalog No. 50931365
Encompass_Preferred
Bath Liquid Therm 240 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 50 to 240oC, 20 L Drum This item is non returnable*
Catalog No. 50-931-365 Supplier Lauda Brinkmann Lp Supplier No. LZB 322
Add to Cart
Add to Cart
Request a Quote
Bath Liquid Therm 240 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 50 to 240oC, 20 L Drum This item is non returnable*