Learn More
Lauda Brinkmann Lp THERM 240 BATH LIQUID 5 L DRUM
Catalog No. 50931363
Encompass_Preferred
Bath Liquid Therm 240 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 50 to 240oC, 5 L Drum This item is non returnable*
Catalog No. 50-931-363 Supplier Lauda Brinkmann Lp Supplier No. LZB 122
Bath Liquid Therm 240 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 50 to 240oC, 5 L Drum This item is non returnable*