Learn More
Thermco Products Inc Lollipop Digital Bottle Thermometer, -50º to 70C
Catalog No. NC1342469
Encompass_Preferred
Lollipop Digital Bottle Thermometers accurately monitors the temperatures of, refrigerators, incubators, block heater/dry baths, and rooms. The minimum/maximum feature monitors critical temperature for any period of time, such as overnight or weekend.
Catalog No. NC1342469 Supplier Thermco Products Inc Supplier No. ACC304INCL
Lollipop Digital Bottle Thermometers accurately monitors the temperatures of, refrigerators, incubators, block heater/dry baths, and rooms. The minimum/maximum feature monitors critical temperature for any period of time, such as overnight or weekend.