Learn More
Cooper Atkins Corporation THERM. DIG.POCKET -40/-450F
Catalog No. NC9446908
Encompass_Preferred
This item has a minimum qty of 5 per supplier requirements.
Our DFP450W Digital Pocket Test with adjustable Temperature Alarm was specifically designed for the harshest foodservice environments. The Instrument can be programmed to provide a visual alarm (blinking display) once the set temperature is reached. The initial default alarm temperature is 140oF (60oC). Guaranteed Accurate for Life means there is no need of field adjusting of calibration settings and no risk of introducing error into the instrument. IPX7 waterproof rated and used to test dishwasher temperatures. Temperature Range: -40 to 450F (-40 to 232C).
Catalog No. NC9446908 Supplier Cooper Atkins Corporation Supplier No. DFP450W08
This item has a minimum qty of 5 per supplier requirements.
Our DFP450W Digital Pocket Test with adjustable Temperature Alarm was specifically designed for the harshest foodservice environments. The Instrument can be programmed to provide a visual alarm (blinking display) once the set temperature is reached. The initial default alarm temperature is 140oF (60oC). Guaranteed Accurate for Life means there is no need of field adjusting of calibration settings and no risk of introducing error into the instrument. IPX7 waterproof rated and used to test dishwasher temperatures. Temperature Range: -40 to 450F (-40 to 232C).