Learn More
Med Vet International Non-Contact Skin Surface Thermometer Veridian infrared Skin Probe Handheld, 09-178, Each
Catalog No. 502828966
Encompass_Preferred
Non-invasive temperature reading of a person or object 1-second readout Measures body and object/liquid temperatures Body temperature measurements taken at forehead Fever alert display icon Fahrenheit/Celsius measurements
Catalog No. 50-282-8966 Supplier Med Vet International Supplier No. 1226096EA
Non-invasive temperature reading of a person or object 1-second readout Measures body and object/liquid temperatures Body temperature measurements taken at forehead Fever alert display icon Fahrenheit/Celsius measurements