Learn More
Med Vet International Non-Contact Skin Surface Thermometer Homedics infrared Skin Probe Handheld, Tie-240, Each
Catalog No. 502832155
Encompass_Preferred
Non-Contact Infrared Technology 3-Color Backlight With High Fever Alarm Clinically Proven Accuracy Measurement In 2 Seconds Deg F/Deg C Options Stores Up To 50 Results In Memory Reads Body, Food, Liquid, And Room Temperature Date Stamp Auto Power-Off For At Home And Professional Use Low Battery Indicator
Catalog No. 50-283-2155 Supplier Med Vet International Supplier No. 1209456EA
Non-Contact Infrared Technology 3-Color Backlight With High Fever Alarm Clinically Proven Accuracy Measurement In 2 Seconds Deg F/Deg C Options Stores Up To 50 Results In Memory Reads Body, Food, Liquid, And Room Temperature Date Stamp Auto Power-Off For At Home And Professional Use Low Battery Indicator