Learn More
Med Vet International Non-Contact Skin Surface Thermometer Veridian infrared Skin Probe Handheld, 09-178, 50/Case
Catalog No. 502829969
Encompass_Preferred
Non-invasive temperature reading of a person or object 1-second readout Measures body and object/liquid temperatures Body temperature measurements taken at forehead Fever alert display icon Fahrenheit/Celsius measurements
Catalog No. 50-282-9969 Supplier Med Vet International Supplier No. 1226096CS
Non-invasive temperature reading of a person or object 1-second readout Measures body and object/liquid temperatures Body temperature measurements taken at forehead Fever alert display icon Fahrenheit/Celsius measurements