Learn More
Med Vet International McKesson Refrigerator / Freezer Thermometer, Digital Display, -58° to +158°F Range, MCK821RFV, Each
Catalog No. 503046369
Encompass_Preferred
Digital Refrigerator / Freezer Thermometer with Alarm McKesson Fahrenheit / Celsius -58° to +158°F (-50° to +70°C) Glycol Bottle Probe Multiple Mounting Options Battery Operated Certified, serialized and traceable and has been tested against a standard calibrated at the National Institute of Standards and Technology (NIST) Temperature-certified to monitor refrigerators or freezers Probe is housed in a 5 mL, unbreakable plastic vial filled with a Bio-Safe Glycol / Water medium and allows internal or external sensor reading display Alarm display with real time and date stamp, low / high limit settings, with visual, flashing LED light and audible beeping indicators Optional display formats for time, date, °C / °F Flip-out desk stand, wall-mounting key hole and magnetic and hook-and-loop attachments allow multiple mounting options
Catalog No. 50-304-6369 Supplier Med Vet International Supplier No. 1074438EA
Digital Refrigerator / Freezer Thermometer with Alarm McKesson Fahrenheit / Celsius -58° to +158°F (-50° to +70°C) Glycol Bottle Probe Multiple Mounting Options Battery Operated Certified, serialized and traceable and has been tested against a standard calibrated at the National Institute of Standards and Technology (NIST) Temperature-certified to monitor refrigerators or freezers Probe is housed in a 5 mL, unbreakable plastic vial filled with a Bio-Safe Glycol / Water medium and allows internal or external sensor reading display Alarm display with real time and date stamp, low / high limit settings, with visual, flashing LED light and audible beeping indicators Optional display formats for time, date, °C / °F Flip-out desk stand, wall-mounting key hole and magnetic and hook-and-loop attachments allow multiple mounting options