Learn More
Ptc Instruments THERM SPOT CHECK 0-525C
Catalog No. NC9063869
Encompass
Direct Contact Spot-Check Thermometer, Temp Range 10 to 525 deg.C, Accuracy +/-2% of full scale range, Dimensions: dia. 1-3/4 in. (4.4cm), Height 1/2 in. (13cm), Weight 1/2 oz.
Catalog No. NC9063869 Supplier Ptc Instruments Supplier No. 573C
Direct Contact Spot-Check Thermometer, Temp Range 10 to 525 deg.C, Accuracy +/-2% of full scale range, Dimensions: dia. 1-3/4 in. (4.4cm), Height 1/2 in. (13cm), Weight 1/2 oz.