Learn More
Grainger Therma Level Gauge HDPE D 23in
Catalog No. 502766882
Encompass_Preferred
The Grainger 16N621 is a Westward brand 1/4-inch drive socket set. It includes 17 sockets ranging from 4mm to 14mm in both standard and deep configurations. The sockets are made from chrome vanadium steel and have a black phosphate finish. The set comes packaged in a molded plastic case for storage and portability. This socket set is ideal for automotive and light industrial applications that require metric sized fasteners. The lifetime warranty provides peace of mind that the sockets are a quality and durable product.
Catalog No. 50-276-6882 Supplier Grainger Supplier No. 16N621
The Grainger 16N621 is a Westward brand 1/4-inch drive socket set. It includes 17 sockets ranging from 4mm to 14mm in both standard and deep configurations. The sockets are made from chrome vanadium steel and have a black phosphate finish. The set comes packaged in a molded plastic case for storage and portability. This socket set is ideal for automotive and light industrial applications that require metric sized fasteners. The lifetime warranty provides peace of mind that the sockets are a quality and durable product.