Learn More
Grainger Therma Level Gauge HDPE D 46in
Catalog No. 502766369
Encompass_Preferred
The Therma Level Gauge HDPE D-46IN is a 46 inch diameter high density polyethylene liquid level gauge made by Therma. It has a maximum temperature rating of 150°F and is compatible with water, mild chemicals, and petroleum products. This gauge has a flanged back and is designed for bulk storage tanks. It features a clear impact-resistant lens for easy viewing of liquid level. The lens is made of scratch resistant polycarbonate. This gauge can withstand up to 5 psi of pressure. It comes with stainless steel mounting hardware. The overall dimensions are 46 x 3 x 46 inches. This American made liquid level gauge is ideal for monitoring liquid levels in large tanks and vessels.
Catalog No. 50-276-6369 Supplier Grainger Supplier No. 16N627
The Therma Level Gauge HDPE D-46IN is a 46 inch diameter high density polyethylene liquid level gauge made by Therma. It has a maximum temperature rating of 150°F and is compatible with water, mild chemicals, and petroleum products. This gauge has a flanged back and is designed for bulk storage tanks. It features a clear impact-resistant lens for easy viewing of liquid level. The lens is made of scratch resistant polycarbonate. This gauge can withstand up to 5 psi of pressure. It comes with stainless steel mounting hardware. The overall dimensions are 46 x 3 x 46 inches. This American made liquid level gauge is ideal for monitoring liquid levels in large tanks and vessels.