Learn More
Grainger Therma Level Gauge HDPE D 57in
Catalog No. 502766761
Encompass_Preferred
The Krueger Therma Level Gauge For 57-16N606 is a liquid level gauge designed for use with Krueger air handlers. It features a clear polycarbonate tube with colored liquid and scale to visually indicate refrigerant level. This gauge has a temperature range of 0°F to 250°F, a working pressure of 435 psi, and 1/4" FFL connections. The tube length is 12" and the overall dimensions are 3-1/2" x 2-1/2" x 12". This OEM replacement gauge allows for quick visual monitoring of refrigerant level in Krueger air handling units.
Catalog No. 50-276-6761 Supplier Grainger Supplier No. 16N632
The Krueger Therma Level Gauge For 57-16N606 is a liquid level gauge designed for use with Krueger air handlers. It features a clear polycarbonate tube with colored liquid and scale to visually indicate refrigerant level. This gauge has a temperature range of 0°F to 250°F, a working pressure of 435 psi, and 1/4" FFL connections. The tube length is 12" and the overall dimensions are 3-1/2" x 2-1/2" x 12". This OEM replacement gauge allows for quick visual monitoring of refrigerant level in Krueger air handling units.