Learn More
Grainger Therma Level Gauge HDPE D 64in
Catalog No. 502764056
Encompass_Preferred
The Krueger Therma Level Gauge For 64 is a liquid-filled thermometer that indicates the level of liquid in a tank. It has a 64 inch long glass tube with gallon increments marked for easy reading. The red liquid inside provides high visibility. It can withstand temperatures from -40 to 250°F. The gauge attaches via a 3/4 inch NPT fitting and requires a 1/2 inch NPT tank connection. This rugged and accurate gauge is ideal for measuring liquid levels in industrial tanks.
Catalog No. 50-276-4056 Supplier Grainger Supplier No. 16N635
Add to Cart
Add to Cart
The Krueger Therma Level Gauge For 64 is a liquid-filled thermometer that indicates the level of liquid in a tank. It has a 64 inch long glass tube with gallon increments marked for easy reading. The red liquid inside provides high visibility. It can withstand temperatures from -40 to 250°F. The gauge attaches via a 3/4 inch NPT fitting and requires a 1/2 inch NPT tank connection. This rugged and accurate gauge is ideal for measuring liquid levels in industrial tanks.