Learn More
Grainger Therma Level Gauge HDPE D 86in
Catalog No. 502764453
Encompass_Preferred
The Krueger Therma Level Gauge is a liquid-filled thermometer designed for use with hot water heating systems to indicate water level and temperature. This gauge has a 4 inch diameter dial with a temperature range of 30 to 240 degrees Fahrenheit, and level indicators from empty to full. The bronze body has 1/2 inch NPT connections and is made in the USA. This gauge allows you to monitor boiler water level and temperature in hydronic heating applications.
Catalog No. 50-276-4453 Supplier Grainger Supplier No. 16N639
The Krueger Therma Level Gauge is a liquid-filled thermometer designed for use with hot water heating systems to indicate water level and temperature. This gauge has a 4 inch diameter dial with a temperature range of 30 to 240 degrees Fahrenheit, and level indicators from empty to full. The bronze body has 1/2 inch NPT connections and is made in the USA. This gauge allows you to monitor boiler water level and temperature in hydronic heating applications.