Learn More
Grainger Therma Level Gauge HDPE D 86in
Catalog No. 502765903
Encompass_Preferred
The Krueger Therma Level Gauge For 86 is a liquid level gauge designed for Krueger HVAC units to indicate refrigerant level. It has a copper float which rises and falls with the refrigerant level. The gauge attaches to the liquid line and provides a clear view of refrigerant level through the sight glass. This universal replacement gauge fits multiple Krueger unit models including Mark VI, DSC, KSSC, and more. It measures 11 inches in length and has 1/4 inch flare connections. The Therma Level Gauge is made in the USA and allows proper monitoring of refrigerant charge.
Catalog No. 50-276-5903 Supplier Grainger Supplier No. 16N612
The Krueger Therma Level Gauge For 86 is a liquid level gauge designed for Krueger HVAC units to indicate refrigerant level. It has a copper float which rises and falls with the refrigerant level. The gauge attaches to the liquid line and provides a clear view of refrigerant level through the sight glass. This universal replacement gauge fits multiple Krueger unit models including Mark VI, DSC, KSSC, and more. It measures 11 inches in length and has 1/4 inch flare connections. The Therma Level Gauge is made in the USA and allows proper monitoring of refrigerant charge.