Learn More
Grainger Therma Level Gauge HDPE D 86in
Catalog No. 502764525
Encompass_Preferred
The Grainger Therma Level Gauge HDPE D-86in is a durable, high-density polyethylene level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. It features a 86" long clear tube with dual scale in inches and centimeters and color-coded red/green sections for quick visual reference. The heavy-duty polypropylene case provides protection. It can withstand temperatures up to 180°F. The gauge attaches via 3/4" NPT male connection and is made in the USA.
Catalog No. 50-276-4525 Supplier Grainger Supplier No. 16N641
The Grainger Therma Level Gauge HDPE D-86in is a durable, high-density polyethylene level gauge designed for use in industrial tanks and vessels to visually indicate liquid level. It features a 86" long clear tube with dual scale in inches and centimeters and color-coded red/green sections for quick visual reference. The heavy-duty polypropylene case provides protection. It can withstand temperatures up to 180°F. The gauge attaches via 3/4" NPT male connection and is made in the USA.