Learn More
Grainger Therma Level Gauge HDPE D 96in
Catalog No. 502764057
Encompass_Preferred
The Grainger Therma Level Gauge HDPE D-96in is a 96 inch long dipstick made of high density polyethylene plastic with markings every 1/8 inch for measuring liquid levels. It has a hermetically sealed bottom to prevent wicking and is suitable for use with water, diesel fuel, solvents, caustics and dilute acids. The gauge has a temperature rating of 140°F and comes with an ABS plastic top cap. It can be used for tanks up to 108 inches deep.
Catalog No. 50-276-4057 Supplier Grainger Supplier No. 16N643
The Grainger Therma Level Gauge HDPE D-96in is a 96 inch long dipstick made of high density polyethylene plastic with markings every 1/8 inch for measuring liquid levels. It has a hermetically sealed bottom to prevent wicking and is suitable for use with water, diesel fuel, solvents, caustics and dilute acids. The gauge has a temperature rating of 140°F and comes with an ABS plastic top cap. It can be used for tanks up to 108 inches deep.