Learn More
Grainger Therma Level Gauge HDPE D 96in
Catalog No. 502764585
Encompass_Preferred
The Krueger Therma Level Gauge For 96 is a liquid-filled thermometer designed for use with Krueger metal coolers and beverage dispensers. It features a 10 inch rigid stem and a 2 inch diameter dial with Fahrenheit readings. The gauge allows monitoring of product temperature and indicates when the metal cooler needs recharged with ice. It has a temperature range of 30 to 100 degrees Fahrenheit. The gauge installs easily by threading into a 1/4 inch NPT coupling on the cooler and includes mounting hardware. It is made in the USA by Krueger.
Catalog No. 50-276-4585 Supplier Grainger Supplier No. 16N645
The Krueger Therma Level Gauge For 96 is a liquid-filled thermometer designed for use with Krueger metal coolers and beverage dispensers. It features a 10 inch rigid stem and a 2 inch diameter dial with Fahrenheit readings. The gauge allows monitoring of product temperature and indicates when the metal cooler needs recharged with ice. It has a temperature range of 30 to 100 degrees Fahrenheit. The gauge installs easily by threading into a 1/4 inch NPT coupling on the cooler and includes mounting hardware. It is made in the USA by Krueger.