Learn More
Grainger Therma Level Gauge HDPE D 96in
Catalog No. 502764596
Encompass_Preferred
The Grainger 16N644 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction and is made of nylon for scrubbing and cleaning. This 6 inch by 9 inch hand pad has a medium duty abrasiveness level. It can be used for a variety of cleaning applications including prep work, finish work, and cleaning equipment. The open web design allows dirt and debris to move through the pad during use.
Catalog No. 50-276-4596 Supplier Grainger Supplier No. 16N644
The Grainger 16N644 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction and is made of nylon for scrubbing and cleaning. This 6 inch by 9 inch hand pad has a medium duty abrasiveness level. It can be used for a variety of cleaning applications including prep work, finish work, and cleaning equipment. The open web design allows dirt and debris to move through the pad during use.