Learn More
Grainger Therma Level Gauge HDPE D 96in
Catalog No. 502766760
Encompass_Preferred
The Grainger 16N614 is a 16 oz. dead blow plastic hammer with a 2-1/2 inch diameter striking face. It has a hollow head filled with steel and lead shot to prevent rebound and absorb shock. The handle is made of fiberglass reinforced nylon for durability. This hammer allows you to strike delicate surfaces without marring and is well-suited for assembling equipment or driving pins and stakes. It provides the heft of steel with the soft striking properties of plastic. The total length is 12-1/2 inches.
Catalog No. 50-276-6760 Supplier Grainger Supplier No. 16N614
Add to Cart
Add to Cart
The Grainger 16N614 is a 16 oz. dead blow plastic hammer with a 2-1/2 inch diameter striking face. It has a hollow head filled with steel and lead shot to prevent rebound and absorb shock. The handle is made of fiberglass reinforced nylon for durability. This hammer allows you to strike delicate surfaces without marring and is well-suited for assembling equipment or driving pins and stakes. It provides the heft of steel with the soft striking properties of plastic. The total length is 12-1/2 inches.