Learn More
Analytical Sales & Services Thermal Adapter Plate for 17P687Z PP Collection plate
Catalog No. 5000806437
Encompass
• For improved temperature transfer to samples\n• For use with 17P687Z (Advantage 1mL 96-Well , Ultra Clear Polypropylene Plate, Round Top, Round Wells)\n\nThe Thermal Adapter Plate conforms to the bottom shape of the collection plate wells to ensure uniform thermal transfer for heating and cooling when used in conjunction with a heating block or chiller (not included). The plate features a SLAS footprint and easy to identify purple anodized coating.\n\nAnalytical Sales & Services can make Thermal Adapter Plates to accommodate select polypropylene collection plates (minimum quantities may apply). Please call 973-616-0700 for more information.
Catalog No. 50-008-06437 Supplier Analytical Sales & Services Supplier No. 17P687ZTAP
• For improved temperature transfer to samples\n• For use with 17P687Z (Advantage 1mL 96-Well , Ultra Clear Polypropylene Plate, Round Top, Round Wells)\n\nThe Thermal Adapter Plate conforms to the bottom shape of the collection plate wells to ensure uniform thermal transfer for heating and cooling when used in conjunction with a heating block or chiller (not included). The plate features a SLAS footprint and easy to identify purple anodized coating.\n\nAnalytical Sales & Services can make Thermal Adapter Plates to accommodate select polypropylene collection plates (minimum quantities may apply). Please call 973-616-0700 for more information.