Learn More
Analytical Sales & Services Thermal Adapter Plate for 1ml TrueTaper Collection Plate
Catalog No. 5000806253
Encompass
• For improved temperature transfer to samples\n• For use with 1mL, 96-well TrueTaper collection (part number 968810 )\n\nThe Thermal Adapter Plate conforms to the bottom shape of the collection plate wells to ensure uniform thermal transfer for heating and cooling when used in conjunction with a heating block or chiller (not included). The plate features a SLAS footprint and easy to identify purple anodized coating.\n\nAnalytical Sales & Services can make Thermal Adapter Plates to accommodate select polypropylene collection plates (minimum quantities may apply). Please call 973-616-0700 for more information.
Catalog No. 50-008-06253 Supplier Analytical Sales & Services Supplier No. 968810TAP
• For improved temperature transfer to samples\n• For use with 1mL, 96-well TrueTaper collection (part number 968810 )\n\nThe Thermal Adapter Plate conforms to the bottom shape of the collection plate wells to ensure uniform thermal transfer for heating and cooling when used in conjunction with a heating block or chiller (not included). The plate features a SLAS footprint and easy to identify purple anodized coating.\n\nAnalytical Sales & Services can make Thermal Adapter Plates to accommodate select polypropylene collection plates (minimum quantities may apply). Please call 973-616-0700 for more information.