Learn More
Analytical Sales & Services Thermal Adapter Plate for 384-well (38200) Collection Plates
Catalog No. 5000804692
Encompass
• For improved temperature transfer to samples\n• For use with 38200 (Advantage 200µL, Polypropylene Square 384 Well Plate, CLEAR)\n• Designed and machined for optimal conformance to the bottom well geometry\n• Ideal if seeking enhanced thermal transfer compared to FLATTAP\n• SLAS dimensions\n• Fits Thermo Ultimate 3000 carousel\n\nThe Thermal Adapter Plate conforms to the bottom shape of the collection plate wells to ensure uniform thermal transfer for heating and cooling when used in conjunction with a heating block or chiller (not included). The plate features a SLAS footprint and easy to identify purple anodized coating.\n\nAnalytical Sales & Services can make Thermal Adapter Plates to accommodate select polypropylene collection plates (minimum quantities may apply). Please call 973-616-0700 for more information.
Catalog No. 50-008-04692 Supplier Analytical Sales & Services Supplier No. 38200TAP
• For improved temperature transfer to samples\n• For use with 38200 (Advantage 200µL, Polypropylene Square 384 Well Plate, CLEAR)\n• Designed and machined for optimal conformance to the bottom well geometry\n• Ideal if seeking enhanced thermal transfer compared to FLATTAP\n• SLAS dimensions\n• Fits Thermo Ultimate 3000 carousel\n\nThe Thermal Adapter Plate conforms to the bottom shape of the collection plate wells to ensure uniform thermal transfer for heating and cooling when used in conjunction with a heating block or chiller (not included). The plate features a SLAS footprint and easy to identify purple anodized coating.\n\nAnalytical Sales & Services can make Thermal Adapter Plates to accommodate select polypropylene collection plates (minimum quantities may apply). Please call 973-616-0700 for more information.