Learn More
Analytical Sales & Services Thermal Adapter Plate for Flat-Bottom Collection Plates
Catalog No. 5000805210
Encompass
Thermal Adapter Plate for Flat-Bottom Collection Plates\n\n• For improved temperature transfer to samples\n• Ideal for use with flat bottom well plates\n• A great generic option for most 384-well plates\n• Works great for many types of flat bottom cell culture plates!\n\nThe Thermal Adapter Plate ensures uniform thermal transfer for heating and cooling when used in conjunction with a heating block or chiller (not included). The plate features a SLAS footprint and easy to identify purple anodized coating.\n\nNote: compatibility has not been determined for all plate types. Some flat bottom plates may not be compatible. Please review the dimensions contained in the public print or contact Analytical Sales to ensure compatibility.\n\nAnalytical Sales & Services can make Thermal Adapter Plates to accommodate select polypropylene collection plates (minimum quantities may apply). Please call 973-616-0700 for more information.
Catalog No. 50-008-05210 Supplier Analytical Sales & Services Supplier No. FLATTAP
Thermal Adapter Plate for Flat-Bottom Collection Plates\n\n• For improved temperature transfer to samples\n• Ideal for use with flat bottom well plates\n• A great generic option for most 384-well plates\n• Works great for many types of flat bottom cell culture plates!\n\nThe Thermal Adapter Plate ensures uniform thermal transfer for heating and cooling when used in conjunction with a heating block or chiller (not included). The plate features a SLAS footprint and easy to identify purple anodized coating.\n\nNote: compatibility has not been determined for all plate types. Some flat bottom plates may not be compatible. Please review the dimensions contained in the public print or contact Analytical Sales to ensure compatibility.\n\nAnalytical Sales & Services can make Thermal Adapter Plates to accommodate select polypropylene collection plates (minimum quantities may apply). Please call 973-616-0700 for more information.