Learn More
Julabo USA Inc 10L Thermal C10 bath fluid -20…+160 °C
SDP
Supplier:  Julabo USA Inc 8891404
Encompass_Preferred
Bath Accessories, Julabo, Thermal C10 - 10 Liter -20°C +160 °C. 10 cSt polydimethylsiloxane, colorless
Catalog No. 50-753-4015