Learn More
Julabo USA Inc 5L Thermal C10 bath fluid -20…+160 °C
SDP
Catalog No. 507534016
Encompass_Preferred
Bath Accessories, Julabo, Thermal C10 - 5 Liter -20°C +160 °C. 10 cSt polydimethylsiloxane, colorless
Catalog No. 50-753-4016 Supplier Julabo USA Inc Supplier No. 8891405
Bath Accessories, Julabo, Thermal C10 - 5 Liter -20°C +160 °C. 10 cSt polydimethylsiloxane, colorless