Learn More
Julabo USA Inc 5L Thermal C2 bath fluid -90…+60 °C
SDP
Catalog No. 507534012
Encompass_Preferred
Bath Accessories, Julabo, Thermal C2 - 5 Liter; -90°C +60 °C. 2 cSt polydimethylsiloxane, colorless
Catalog No. 50-753-4012 Supplier Julabo USA Inc Supplier No. 8891401
Bath Accessories, Julabo, Thermal C2 - 5 Liter; -90°C +60 °C. 2 cSt polydimethylsiloxane, colorless