Learn More
Julabo USA Inc 10L Thermal C20S bath fluid 0…+220 °C
SDP
Catalog No. 507534019
Encompass_Preferred
Bath Accessories, Julabo, Thermal C20S - 10 Liter; 0°C +220 °C. 20 cSt stabilized polydimethylsiloxane, light brown.
Catalog No. 50-753-4019 Supplier Julabo USA Inc Supplier No. 8891408
Bath Accessories, Julabo, Thermal C20S - 10 Liter; 0°C +220 °C. 20 cSt stabilized polydimethylsiloxane, light brown.