Learn More
Julabo USA Inc 5L Thermal C20S bath fluid 0…+220 °C
SDP
Catalog No. 507534020
Encompass_Preferred
Bath Accessories, Julabo, Thermal C20S - 5 Liter; 0°C +220 °C. 20 cSt stabilized polydimethylsiloxane, light brown.
Catalog No. 50-753-4020 Supplier Julabo USA Inc Supplier No. 8891409
Bath Accessories, Julabo, Thermal C20S - 5 Liter; 0°C +220 °C. 20 cSt stabilized polydimethylsiloxane, light brown.