Learn More
Julabo USA Inc 10L Thermal C5 bath fluid -60…+110 °C
SDP
Catalog No. 507534013
Encompass_Preferred
Bath Accessories, Julabo, Thermal C5 - 10 Liter -60°C +90 °C. 5 cSt polydimethylsiloxane, colorless
Catalog No. 50-753-4013 Supplier Julabo USA Inc Supplier No. 8891402
Bath Accessories, Julabo, Thermal C5 - 10 Liter -60°C +90 °C. 5 cSt polydimethylsiloxane, colorless