Learn More
Julabo USA Inc 5L Thermal C5 bath fluid -60…+110 °C
SDP
Catalog No. 507534014
Encompass_Preferred
Bath Accessories, Julabo, Thermal C5 - 5 Liter -60°C +90 °C. 5 cSt polydimethylsiloxane, colorless
Catalog No. 50-753-4014 Supplier Julabo USA Inc Supplier No. 8891403
Bath Accessories, Julabo, Thermal C5 - 5 Liter -60°C +90 °C. 5 cSt polydimethylsiloxane, colorless