Learn More
Julabo USA Inc 10L Thermal C50S bath fluid +20…+250 °C
SDP
Catalog No. 507534021
Encompass_Preferred
Bath Accessories, Julabo, Thermal C50S - 10 Liter; +20°C +250 °C. 50 cSt stabilized polydimethylsiloxane, light brown.
Catalog No. 50-753-4021 Supplier Julabo USA Inc Supplier No. 8891410
Bath Accessories, Julabo, Thermal C50S - 10 Liter; +20°C +250 °C. 50 cSt stabilized polydimethylsiloxane, light brown.