Learn More
Julabo USA Inc 5L Thermal C50S bath fluid +20…+250 °C
SDP
Catalog No. 507534022
Encompass_Preferred
Bath Accessories, Julabo, Thermal C50S - 5 Liter; +20°C +250 °C. 50 cSt stabilized polydimethylsiloxane, light brown.
Catalog No. 50-753-4022 Supplier Julabo USA Inc Supplier No. 8891411
Bath Accessories, Julabo, Thermal C50S - 5 Liter; +20°C +250 °C. 50 cSt stabilized polydimethylsiloxane, light brown.